AMBH303C66C4 Ambeed, Inc.

2-(1H-Indol-1-yl)ethanol

  • MDL number MFCD12964923
  • Cas Number : 121459-15-2
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/121459-15-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/121459-15-2.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart