AMBH303C66C6 Ambeed, Inc.

2-(Trifluoromethyl)pyrimidine

  • MDL number MFCD10698646
  • Cas Number : 116470-67-8
  • Molecular Weight : 148.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point -4.5
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 69�t 760 mmHg
Product Link https://www.ambeed.com/products/116470-67-8.html

Safety Information

signalword Danger
pcodes P210-P305+P351+P338
hcodes 3
MSDS https://www.ambeed.com/products/116470-67-8.html
pictograms GHS02,GHS07
hstatements H225-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart