AMBH303C66DA Ambeed, Inc.

2,6-Dichloro-3-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD11109540
  • Cas Number : 276702-53-5
  • Molecular Weight : 290.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/276702-53-5.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/276702-53-5.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart