AMBH303C66E1 Ambeed, Inc.

Methyl 2-(5-hydroxy-2,3-dihydro-1H-inden-1-yl)acetate

  • MDL number MFCD11100971
  • Cas Number : 856169-08-9
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/856169-08-9.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/856169-08-9.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart