AMBH303C66E9 Ambeed, Inc.

N-(4,6-Dichloropyrimidin-5-yl)formamide

  • MDL number MFCD04974511
  • Cas Number : 123240-66-4
  • Molecular Weight : 192.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/123240-66-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/123240-66-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart