AMBH303C66EB Ambeed, Inc.

Ro 10-5824 2HCl

  • MDL number MFCD08703115
  • Cas Number : 189744-94-3
  • Molecular Weight : 353.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Methyl-5-((4-phenyl-5,6-dihydropyridin-1(2H)-yl)methyl)pyrimidin-4-amine dihydrochloride
Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/189744-94-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/189744-94-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart