AMBH303C66F4 Ambeed, Inc.

(S)-1-(3-Chloro-4-methylphenyl)ethanamine hydrochloride

  • MDL number MFCD16294091
  • Cas Number : 1269469-78-4
  • Molecular Weight : 206.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1269469-78-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1269469-78-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart