AMBH303C6725 Ambeed, Inc.

2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine

  • MDL number MFCD29047055
  • Cas Number : 77989-15-2
  • Molecular Weight : 388.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-146�t N/A mmHg
Purity 99
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/77989-15-2.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
MSDS https://www.ambeed.com/products/77989-15-2.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart