AMBH303C672D Ambeed, Inc.

3-(Trifluoromethoxy)pyridin-4-amine

  • MDL number MFCD19690275
  • Cas Number : 827586-90-3
  • Molecular Weight : 178.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/827586-90-3.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/827586-90-3.html
pictograms GHS07,GHS05
hstatements H302-H318
Availability details for
Quantity
  • Availability
  • Add to Cart