AMBH303C6756 Ambeed, Inc.

tert-Butyl 3-cyano-4-hydroxypyrrolidine-1-carboxylate

  • MDL number MFCD01318457
  • Cas Number : 197143-33-2
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-96
Purity 96
Flash Point 172
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 361�t 760 mmHg
Product Link https://www.ambeed.com/products/197143-33-2.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/197143-33-2.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart