AMBH303C677F Ambeed, Inc.

2-(Chloromethyl)-5-methylthieno[2,3-d]pyrimidin-4(3H)-one

  • MDL number MFCD04115397
  • Cas Number : 568577-81-1
  • Molecular Weight : 214.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/568577-81-1.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/568577-81-1.html
pictograms GHS07
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart