AMBH303C6788 Ambeed, Inc.

3-Isopropylpiperazine-2,5-dione

  • MDL number MFCD00480885
  • Cas Number : 14771-77-8
  • Molecular Weight : 156.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 280�t N/A mmHg
Purity 97
Flash Point 216.5
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 445.7�t 760 mmHg
Product Link https://www.ambeed.com/products/14771-77-8.html

Safety Information

signalword Warning
pcodes P261-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/14771-77-8.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart