AMBH303C678C Ambeed, Inc.

Methyl 2-acetylbenzoate

  • MDL number MFCD20485857
  • Cas Number : 1077-79-8
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1077-79-8.html

Safety Information

signalword Warning
pcodes P261-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/1077-79-8.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart