AMBH303C67C6 Ambeed, Inc.

5-Bromo-2,3-dihydro-1H-pyrrolo[3,2-b]pyridine

  • MDL number MFCD18250826
  • Cas Number : 1260671-35-9
  • Molecular Weight : 199.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1260671-35-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1260671-35-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart