AMBH303C67F7 Ambeed, Inc.

Sodium 4-Amino-1-naphthalenesulfonate Tetrahydrate

  • MDL number MFCD00044844
  • Cas Number : 130-13-2
  • Molecular Weight : 245.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/130-13-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/130-13-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart