AMBH303C6817 Ambeed, Inc.

2-(4-Amino-3-chlorophenyl)butanoic acid

  • MDL number MFCD24487840
  • Cas Number : 25814-13-5
  • Molecular Weight : 213.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/25814-13-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/25814-13-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart