AMBH303C6876 Ambeed, Inc.

2,4-Di([1,1'-biphenyl]-4-yl)-6-chloro-1,3,5-triazine

  • MDL number MFCD27979765
  • Cas Number : 182918-13-4
  • Molecular Weight : 419.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 370.6
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 649.6�0�t 760 mmHg
Product Link https://www.ambeed.com/products/182918-13-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/182918-13-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart