AMBH303C68E8 Ambeed, Inc.

(1,3,5-Trimethyl-1H-pyrazol-4-yl)methanamine

  • MDL number MFCD02055811
  • Cas Number : 352018-93-0
  • Molecular Weight : 139.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/352018-93-0.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/352018-93-0.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart