AMBH303C68EF Ambeed, Inc.

Ethyl 3-(3,4-difluorophenyl)-5-methyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD31728058
  • Cas Number : 2288710-15-4
  • Molecular Weight : 265.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/2288710-15-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2288710-15-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart