AMBH303C693C Ambeed, Inc.

Jatrorrhizine

  • MDL number MFCD01310242
  • Cas Number : 3621-38-3
  • Molecular Weight : 338.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/3621-38-3.html

Safety Information

signalword Danger
pcodes P273
MSDS https://www.ambeed.com/products/3621-38-3.html
pictograms GHS09,GHS07
hstatements H302-H400
Availability details for
Quantity
  • Availability
  • Add to Cart