AMBH303C6959 Ambeed, Inc.

tert-Butyl 3-bromo-2,4-dioxopiperidine-1-carboxylate

  • MDL number MFCD24471390
  • Cas Number : 1312412-87-5
  • Molecular Weight : 292.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1312412-87-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1312412-87-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart