AMBH303C6963 Ambeed, Inc.

Ethyl hex-2-enoate

  • MDL number MFCD00036541
  • Cas Number : 27829-72-7
  • Molecular Weight : 142.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/27829-72-7.html

Safety Information

signalword Danger
pcodes P210-P280-P370+P378-P403+P235-P501
MSDS https://www.ambeed.com/products/27829-72-7.html
pictograms GHS02
hstatements H227
Availability details for
Quantity
  • Availability
  • Add to Cart