AMBH303C69A5 Ambeed, Inc.

4-(Trifluoromethyl)indoline

  • MDL number MFCD08544326
  • Cas Number : 905274-07-9
  • Molecular Weight : 187.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 208.6�t 760 mmHg
Product Link https://www.ambeed.com/products/905274-07-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/905274-07-9.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart