AMBH303C69A8 Ambeed, Inc.

3-Amino-4-nitrobenzonitrile

  • MDL number MFCD18389919
  • Cas Number : 99512-10-4
  • Molecular Weight : 163.1335

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/99512-10-4.html

Safety Information

signalword Danger
pcodes P261-P280-P302+P352
MSDS https://www.ambeed.com/products/99512-10-4.html
pictograms GHS06
hstatements H302-H311-H317
Availability details for
Quantity
  • Availability
  • Add to Cart