AMBH303C69B4 Ambeed, Inc.

INCB-057643

  • MDL number MFCD31657400
  • Cas Number : 1820889-23-3
  • Molecular Weight : 415.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2,4-Trimethyl-8-(6-methyl-7-oxo-6,7-dihydro-1H-pyrrolo[2,3-c]pyridin-4-yl)-6-(methylsulfonyl)-2H-benzo[b][1,4]oxazin-3(4H)-one
Purity 99
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1820889-23-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1820889-23-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart