AMBH303C69D6 Ambeed, Inc.

Suprofen

  • MDL number MFCD00079572
  • Cas Number : 40828-46-4
  • Molecular Weight : 260.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(4-(Thiophene-2-carbonyl)phenyl)propanoic acid
Purity 98
Flash Point 221.5
Storage temp. Keep in dark place,Sealed in dry,Store in freezer, under -20
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Boiling point 442.6�t 760 mmHg
Product Link https://www.ambeed.com/products/40828-46-4.html

Safety Information

signalword Danger
pcodes P301+P310
hcodes 6.1
MSDS https://www.ambeed.com/products/40828-46-4.html
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart