AMBH303C69E6 Ambeed, Inc.

ETC-1002

  • MDL number N/A
  • Cas Number : 738606-46-7
  • Molecular Weight : 344.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 8-Hydroxy-2,2,14,14-tetramethylpentadecanedioic acid
Purity 98
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/738606-46-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/738606-46-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart