AMBH303C69EF Ambeed, Inc.

5,7-Dichloro-[1,2,4]triazolo[1,5-a]pyridin-2-amine

  • MDL number MFCD18642394
  • Cas Number : 1124383-04-5
  • Molecular Weight : 203.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1124383-04-5.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/1124383-04-5.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart