AMBH303C6A07 Ambeed, Inc.

2-(6-Chloropyridin-2-yl)acetonitrile

  • MDL number MFCD09743542
  • Cas Number : 75279-60-6
  • Molecular Weight : 152.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/75279-60-6.html

Safety Information

signalword Danger
pcodes P261-P280-P301+P310-P302+P350-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/75279-60-6.html
pictograms GHS06
hstatements H301-H310-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart