AMBH303C6A0C Ambeed, Inc.

1-(2-Bromo-4-methylthiazol-5-yl)ethanone

  • MDL number MFCD19370713
  • Cas Number : 1093106-54-7
  • Molecular Weight : 220.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1093106-54-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1093106-54-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart