AMBH303C6A29 Ambeed, Inc.

Chloromethyl isopropyl carbonate

  • MDL number MFCD07375443
  • Cas Number : 35180-01-9
  • Molecular Weight : 152.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 50
Storage temp. Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 147.5�t 760 mmHg
Product Link https://www.ambeed.com/products/35180-01-9.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
hcodes 3,8
MSDS https://www.ambeed.com/products/35180-01-9.html
pictograms GHS02,GHS05
hstatements H226-H314
Availability details for
Quantity
  • Availability
  • Add to Cart