AMBH303C6A2E Ambeed, Inc.

2-((tert-Butoxycarbonyl)amino)-4-methoxy-4-oxobutanoic acid

  • MDL number MFCD16619508
  • Cas Number : 856417-64-6
  • Molecular Weight : 247.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/856417-64-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/856417-64-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart