AMBH303C6A47 Ambeed, Inc.

4-Chloro-8-fluoro-2-(trifluoromethyl)quinazoline

  • MDL number MFCD09954903
  • Cas Number : 959238-18-7
  • Molecular Weight : 250.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/959238-18-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/959238-18-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart