AMBH303C6A4A Ambeed, Inc.

2-Amino-1H-imidazole-5-carboxylic acid

  • MDL number MFCD08668249
  • Cas Number : 860011-60-5
  • Molecular Weight : 127.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/860011-60-5.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/860011-60-5.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart