AMBH303C6A66 Ambeed, Inc.

Tandutinib

  • MDL number MFCD09954147
  • Cas Number : 387867-13-2
  • Molecular Weight : 562.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177-178�t N/A mmHg
Synonym N-(4-Isopropoxyphenyl)-4-(6-methoxy-7-(3-(piperidin-1-yl)propoxy)quinazolin-4-yl)piperazine-1-carboxamide
Purity 98
Flash Point 419.2
Storage temp. Sealed in dry,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 769.5�t 760 mmHg
Product Link https://www.ambeed.com/products/387867-13-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/387867-13-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart