AMBH303C6A7E Ambeed, Inc.

4-(Bromomethyl)-2-fluoro-1-methoxybenzene

  • MDL number MFCD00671768
  • Cas Number : 331-61-3
  • Molecular Weight : 219.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/331-61-3.html

Safety Information

signalword Warning
pcodes P273
MSDS https://www.ambeed.com/products/331-61-3.html
pictograms GHS07
hstatements H302-H412
Availability details for
Quantity
  • Availability
  • Add to Cart