AMBH303C6A97 Ambeed, Inc.

2-(Methoxymethyl)azetidine

  • MDL number MFCD23105901
  • Cas Number : 1290136-94-5
  • Molecular Weight : 101.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 31.3
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Boiling point 114.2�0�t N/A mmHg
Product Link https://www.ambeed.com/products/1290136-94-5.html

Safety Information

signalword Danger
pcodes P261-P305+P351+P338
hcodes 3
MSDS https://www.ambeed.com/products/1290136-94-5.html
pictograms GHS02,GHS07
hstatements H226-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart