AMBH303C6AA7 Ambeed, Inc.

ML 213

  • MDL number MFCD00848936
  • Cas Number : 489402-47-3
  • Molecular Weight : 257.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Mesitylbicyclo[2.2.1]heptane-2-carboxamide
Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/489402-47-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/489402-47-3.html
pictograms GHS07
hstatements H315-H319-H335-H413
Availability details for
Quantity
  • Availability
  • Add to Cart