AMBH303C6AAD Ambeed, Inc.

Potassium hexadecyl hydrogenphosphate

  • MDL number MFCD04112600
  • Cas Number : 19035-79-1
  • Molecular Weight : 360.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/19035-79-1.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/19035-79-1.html
pictograms GHS05
hstatements H318
Availability details for
Quantity
  • Availability
  • Add to Cart