AMBH303C6AB2 Ambeed, Inc.

8-Bromoimidazo[1,2-a]pyrazine

  • MDL number MFCD18072715
  • Cas Number : 69214-34-2
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/69214-34-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/69214-34-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart