AMBH303C6AE6 Ambeed, Inc.

1-(tert-Butyldimethylsilyl)prop-2-en-1-ol

  • MDL number MFCD19687250
  • Cas Number : 105875-75-0
  • Molecular Weight : 172.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 33
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 120�t 760 mmHg
Product Link https://www.ambeed.com/products/105875-75-0.html

Safety Information

signalword Danger
pcodes P210-P403+P235
hcodes 3
MSDS https://www.ambeed.com/products/105875-75-0.html
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart