AMBH303C6B0B Ambeed, Inc.

AF64394

  • MDL number MFCD28411712
  • Cas Number : 1637300-25-4
  • Molecular Weight : 393.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1637300-25-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1637300-25-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart