AMBH303C6B0F Ambeed, Inc.

5-Bromo-2-(bromomethyl)-1,3-difluorobenzene

  • MDL number MFCD03094462
  • Cas Number : 162744-60-7
  • Molecular Weight : 285.9100

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/162744-60-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/162744-60-7.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart