AMBH303C6B1B Ambeed, Inc.

1H-Pyrrolo[3,2-c]pyridine-6-carbonitrile

  • MDL number MFCD11845528
  • Cas Number : 1082040-98-9
  • Molecular Weight : 143.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1082040-98-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1082040-98-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart