AMBH303C6B4F Ambeed, Inc.

GNE-6776

  • MDL number MFCD31657364
  • Cas Number : 2009273-71-4
  • Molecular Weight : 348.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6'-Amino-4'-ethyl-5'-(4-hydroxyphenyl)-N-methyl-[3,3'-bipyridine]-6-carboxamide
Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/2009273-71-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2009273-71-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart