AMBH303C6B7C Ambeed, Inc.

NS 1738

  • MDL number MFCD11100189
  • Cas Number : 501684-93-1
  • Molecular Weight : 365.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/501684-93-1.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/501684-93-1.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart