AMBH303C6BB7 Ambeed, Inc.

1-Iodo-4-isopropyl-2-methoxybenzene

  • MDL number MFCD29096400
  • Cas Number : 1240304-64-6
  • Molecular Weight : 276.1141

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1240304-64-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1240304-64-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart