AMBH31555DBE Ambeed, Inc.

Itttdqp

  • MDL number N/A
  • Cas Number : 1375279-77-8
  • Molecular Weight : 761.90

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1375279-77-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1375279-77-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart