AMBH3155F8E9 Ambeed, Inc.

2-Bromo-1-(3-bromo-2-fluorophenyl)ethanone

  • MDL number MFCD16140216
  • Cas Number : 1204333-47-0
  • Molecular Weight : 295.9300

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form White to yellow powder or crystals or liquid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1204333-47-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1204333-47-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart