AMBH3155F8F7 Ambeed, Inc.

5-Fluoro-2-nitropyridin-3-ol

  • MDL number MFCD11878539
  • Cas Number : 847902-56-1
  • Molecular Weight : 158.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-fluoro-2-nitro-3-pyridinol
Inchi Code 1S/C5H3FN2O3/c6-3-1-4(9)5(7-2-3)8(10)11/h1-2,9H
Inchi Key XCYLKKXQKCXJAE-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/847902-56-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/847902-56-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart